Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Chloroacetyl-DL-norleucine 99.0+%, TCI America™
SDP

Catalog No. C01021G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C01021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C01021G Supplier TCI America Supplier No. C01021G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67206-26-2
Molecular Formula C8H14ClNO3
Molecular Weight (g/mol) 207.65
MDL Number MFCD00059650
InChI Key NTOGPWIZAGBRID-UHFFFAOYNA-N
PubChem CID 306113
IUPAC Name 2-(2-chloroacetamido)hexanoic acid
SMILES CCCCC(NC(=O)CCl)C(O)=O

Specifications

Melting Point 113°C
Color White
Quantity 1 g
Formula Weight 207.65
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Chloroacetyl-DL-norleucine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.