Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Chloroacetyl-DL-valine 98.0+%, TCI America™
SDP

Catalog No. C01061G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C01061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C01061G Supplier TCI America Supplier No. C01061G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4090-17-9
Molecular Formula C7H12ClNO3
Molecular Weight (g/mol) 193.63
MDL Number MFCD00062945
InChI Key LJRISAYPKJORFZ-UHFFFAOYNA-N
PubChem CID 101112
IUPAC Name 2-(2-chloroacetamido)-3-methylbutanoic acid
SMILES CC(C)C(NC(=O)CCl)C(O)=O

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 1 g
Formula Weight 193.63
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Chloroacetyl-DL-valine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.