Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Chloroacetyl-L-valine 98.0+%, TCI America™
SDP

Catalog No. C0107100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0107100MG 100 mg
C01071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0107100MG Supplier TCI America Supplier No. C0107100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2279-16-5
Molecular Formula C7H12ClNO3
Molecular Weight (g/mol) 193.63
MDL Number MFCD00045273
InChI Key LJRISAYPKJORFZ-UHFFFAOYNA-N
PubChem CID 737074
IUPAC Name 2-(2-chloroacetamido)-3-methylbutanoic acid
SMILES CC(C)C(NC(=O)CCl)C(O)=O

Specifications

Melting Point 115°C
Color White
Quantity 100 mg
Formula Weight 193.63
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Chloroacetyl-L-valine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.