Learn More
Medchemexpress LLC N-chlorosuccinimide | 128-09-6 | MFCD00005511 | 99.0% | 133.53 g/mol | C4H4ClNO2 | 500g
SDP
Catalog No. 5000268885
Encompass_Preferred
N-Chlorosuccinimide is a five-membered N-containing heterocyclic molecule N-Chlorosuccinimide is a versatile reagent and can be used for different types of reactions such as aliphatic and aromatic chlorination N-Chlorosuccinimide can enhance the antimicrobial effect of Benzalkonium chloride (HY-B2232) on biofilm Pseudomonas aeruginosa[1][2]
Catalog No. 50-002-68885 Supplier Medchemexpress LLC Supplier No. HYY0532500G
N-Chlorosuccinimide is a five-membered N-containing heterocyclic molecule N-Chlorosuccinimide is a versatile reagent and can be used for different types of reactions such as aliphatic and aromatic chlorination N-Chlorosuccinimide can enhance the antimicrobial effect of Benzalkonium chloride (HY-B2232) on biofilm Pseudomonas aeruginosa[1][2]