Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Cinnamoyl-N-(2,3-xylyl)hydroxylamine 98.0+%, TCI America™
SDP

Catalog No. C10585G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10585G 5 g
C10581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C10585G Supplier TCI America Supplier No. C10585G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 69891-38-9
Molecular Formula C17H17NO2
Molecular Weight (g/mol) 267.328
MDL Number MFCD00059308
InChI Key SLIQYZYALKWWAT-UHFFFAOYSA-N
Synonym N-(2,3-Xylyl)-N-cinnamoylhydroxylamine
PubChem CID 53396032
IUPAC Name N-(2,3-dimethylphenyl)-N-hydroxy-3-phenylprop-2-enamide
SMILES CC1=C(C(=CC=C1)N(C(=O)C=CC2=CC=CC=C2)O)C

Specifications

Melting Point 152°C
Color White
Quantity 5 g
Formula Weight 267.33
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Cinnamoyl-N-(2,3-xylyl)hydroxylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.