Learn More
Medchemexpress LLC N-(cis-3-(methyl(7H-pyrrolo(2,3-d)pyrimidin-4-yl)amino)cyclobutyl)-1-propanesulfonamide | 1622902-68-4 | MFCD30187577 | >98.0% | 323.41 | C14H21N5O2S | 100 MG
SDP
Catalog No. 5000239835
Encompass_Preferred
Abrocitinib is a potent, orally active, and selective JAK1 inhibitor supplied as a research-grade analytical standard. It is provided as a purified solid suitable for in vitro pharmacology, assay development, and analytical reference. Chemical identity: N-(cis-3-(methyl(7H-pyrrolo(2,3-d)pyrimidin-4-yl)amino)cyclobutyl)-1-propanesulfonamide; CAS 1622902-68-4; formula C14H21N5O2S; molecular weight 323.41 g/mol.
  • Potent, selective JAK1 inhibitor for mechanistic and screening studies.
  • High purity (>98.0%) suitable for analytical and assay use.
  • Solid powder form for accurate weighing and formulation.
  • Well-characterized chemical identity with CAS 1622902-68-4.
  • Appropriate for in vitro pharmacology and compound profiling.
Catalog No. 50-002-39835 Supplier Medchemexpress LLC Supplier No. HY107429100MG
Abrocitinib is a potent, orally active, and selective JAK1 inhibitor supplied as a research-grade analytical standard. It is provided as a purified solid suitable for in vitro pharmacology, assay development, and analytical reference. Chemical identity: N-(cis-3-(methyl(7H-pyrrolo(2,3-d)pyrimidin-4-yl)amino)cyclobutyl)-1-propanesulfonamide; CAS 1622902-68-4; formula C14H21N5O2S; molecular weight 323.41 g/mol.
  • Potent, selective JAK1 inhibitor for mechanistic and screening studies.
  • High purity (>98.0%) suitable for analytical and assay use.
  • Solid powder form for accurate weighing and formulation.
  • Well-characterized chemical identity with CAS 1622902-68-4.
  • Appropriate for in vitro pharmacology and compound profiling.