Learn More
Medchemexpress LLC N-cyano-4-dimethylaminopyridinium tetrafluoroborate | 59016-56-7 | 98.0% | 234.99 | 250 MG
SDP
Catalog No. 5000366301
Encompass_Preferred
CDAP is an organic cyanylating reagent used for preparing solid-phase affinity reagents, modifying protein thiols, and delivering nucleotides. It also activates polysaccharides with various chemical properties and can be used to create conjugate vaccines and other immune reagents.
  • Prepares solid-phase affinity reagents.
  • Modifies protein thiols.
  • Delivers nucleotides.
  • Activates polysaccharides with various chemical properties.
  • Creates conjugate vaccines and other immune reagents.
  • Activates a wide range of acidic, neutral, and other classes of polysaccharides.
  • Allows direct conjugation of proteins to activated polysaccharides without additional spacer molecules.
  • Simplifies the preparation of conjugate vaccines.
Catalog No. 50-003-66301 Supplier Medchemexpress LLC Supplier No. 250MGHYW012184
CDAP is an organic cyanylating reagent used for preparing solid-phase affinity reagents, modifying protein thiols, and delivering nucleotides. It also activates polysaccharides with various chemical properties and can be used to create conjugate vaccines and other immune reagents.
  • Prepares solid-phase affinity reagents.
  • Modifies protein thiols.
  • Delivers nucleotides.
  • Activates polysaccharides with various chemical properties.
  • Creates conjugate vaccines and other immune reagents.
  • Activates a wide range of acidic, neutral, and other classes of polysaccharides.
  • Allows direct conjugation of proteins to activated polysaccharides without additional spacer molecules.
  • Simplifies the preparation of conjugate vaccines.