Learn More
Medchemexpress LLC N-cyclobutyl-6-[2-(3-fluorophenyl)ethynyl]-3-pyridinecarboxamide | 1310012-12-4 | 98.1% | 50 MG
SDP
Catalog No. 5000234369
Encompass_Preferred
VU0360172 is a potent and selective positive allosteric modulator of the metabotropic glutamate receptor 5 (mGlu5) used as a research reagent for cellular and in vivo pharmacology studies.
  • Potent mGlu5 positive allosteric modulation (EC50 16 nM).
  • Binding affinity to mGlu5 (Ki 195 nM).
  • High purity suitable for research (98.1%).
  • Molecular formula C18H15FN2O and molecular weight 294.32 g/mol.
  • Solid, light yellow to brown research material supplied as 50 MG quantities.
Catalog No. 50-002-34369 Supplier Medchemexpress LLC Supplier No. HY12058950MG
VU0360172 is a potent and selective positive allosteric modulator of the metabotropic glutamate receptor 5 (mGlu5) used as a research reagent for cellular and in vivo pharmacology studies.
  • Potent mGlu5 positive allosteric modulation (EC50 16 nM).
  • Binding affinity to mGlu5 (Ki 195 nM).
  • High purity suitable for research (98.1%).
  • Molecular formula C18H15FN2O and molecular weight 294.32 g/mol.
  • Solid, light yellow to brown research material supplied as 50 MG quantities.