Learn More
Medchemexpress LLC N-cyclobutyl-6-[2-(3-fluorophenyl)ethynyl]-3-pyridinecarboxamide, monohydrochloride | 1309976-62-2 | 98.9% | 330.78 g·mol⁻¹ | C18H16ClFN2O | 50 MG
SDP
Catalog No. 5000241376
Encompass_Preferred
VU0360172 hydrochloride is a research-grade small molecule and a potent, selective positive allosteric modulator of the metabotropic glutamate receptor 5 (mGlu5). Supplied as a solid for in vitro and in vivo pharmacology studies, it is characterized by nanomolar potency and high chemical purity.
  • Selective mGlu5 PAM with EC50 = 16 nM and Ki = 195 nM.
  • High purity suitable for research use (≈98.9%).
  • Solid form, convenient for storage and formulation.
  • Molecular weight 330.78 g·mol⁻¹; formula C18H16ClFN2O.
  • Recommended sealed storage away from moisture at low temperature.
Catalog No. 50-002-41376 Supplier Medchemexpress LLC Supplier No. HY10750450MG
VU0360172 hydrochloride is a research-grade small molecule and a potent, selective positive allosteric modulator of the metabotropic glutamate receptor 5 (mGlu5). Supplied as a solid for in vitro and in vivo pharmacology studies, it is characterized by nanomolar potency and high chemical purity.
  • Selective mGlu5 PAM with EC50 = 16 nM and Ki = 195 nM.
  • High purity suitable for research use (≈98.9%).
  • Solid form, convenient for storage and formulation.
  • Molecular weight 330.78 g·mol⁻¹; formula C18H16ClFN2O.
  • Recommended sealed storage away from moisture at low temperature.