Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-cyclobutyl-6-[2-(3-fluorophenyl)ethynyl]pyridine-3-carboxamide | 1310012-12-4 | 98.1% | 25 MG
SDP

Catalog No. 5000232177
Encompass_Preferred

VU0360172 is a potent, selective metabotropic glutamate receptor 5 (mGlu5) positive allosteric modulator supplied as a high-purity research reagent. It exhibits low-nanomolar cellular activity and contains an alkyne group for click chemistry applications.

  • Potent mGlu5 positive allosteric modulator; EC50 16 nM in HEK293 cells.
  • High purity (98.07%) solid, suitable for in vitro and in vivo studies.
  • Soluble in DMSO (83.33 mg/mL) and formulatable for in vivo dosing (2.08 mg/mL in 10% DMSO/90% corn oil).
  • Store at 4°C protected from light; in solvent store at -80°C for long-term stability.

Catalog No. 50-002-32177 Supplier Medchemexpress LLC Supplier No. HY12058925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VU0360172 is a potent, selective metabotropic glutamate receptor 5 (mGlu5) positive allosteric modulator supplied as a high-purity research reagent. It exhibits low-nanomolar cellular activity and contains an alkyne group for click chemistry applications.

  • Potent mGlu5 positive allosteric modulator; EC50 16 nM in HEK293 cells.
  • High purity (98.07%) solid, suitable for in vitro and in vivo studies.
  • Soluble in DMSO (83.33 mg/mL) and formulatable for in vivo dosing (2.08 mg/mL in 10% DMSO/90% corn oil).
  • Store at 4°C protected from light; in solvent store at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.