Learn More
Medchemexpress LLC N-cyclobutyl-N-(4-ethylphenyl)-4-hydroxy-2-(oxan-4-yl)-3,4-dihydro-2H-chromene-6-sulfonamide | 2230779-18-5 | 99.3% | C26H33NO5S | 10MG
SDP
Catalog No. 5000258715
Encompass_Preferred
RORγt inverse agonist 23 is a potent, selective, orally available small-molecule inverse agonist of the retinoic acid receptor-related orphan receptor γt (RORγt), intended for preclinical research into RORγt-mediated immune signaling. The compound is provided as a characterized solid with associated structural identifiers to support reproducible research.
  • Potent, selective inverse agonist of RORγt.
  • Orally available small-molecule suitable for in vivo and in vitro studies.
  • High purity (~99.3%).
  • Multiple pack sizes including mg-scale and DMSO solutions.
  • Supplied with structural identifiers for unambiguous identification.
Catalog No. 50-002-58715 Supplier Medchemexpress LLC Supplier No. HY13984710MG
RORγt inverse agonist 23 is a potent, selective, orally available small-molecule inverse agonist of the retinoic acid receptor-related orphan receptor γt (RORγt), intended for preclinical research into RORγt-mediated immune signaling. The compound is provided as a characterized solid with associated structural identifiers to support reproducible research.
  • Potent, selective inverse agonist of RORγt.
  • Orally available small-molecule suitable for in vivo and in vitro studies.
  • High purity (~99.3%).
  • Multiple pack sizes including mg-scale and DMSO solutions.
  • Supplied with structural identifiers for unambiguous identification.