Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-cycloheptyl-6,7-dimethoxy-2-(4-piperidin-1-ylpiperidin-1-yl)quinazolin-4-amine | 864289-85-0 | 100.0% | 467.65 g/mol | C27H41N5O2 | 10 MG
SDP

Catalog No. 5000245134
Encompass_Preferred

C-021 is a small-molecule CCR4 antagonist (CAS 864289-85-0) provided as a high-purity solid for research use. It is intended for in vitro and in vivo pharmacology studies where CCR4 inhibition is required. Supplied quantity: 10 mg; follow recommended solvent and storage guidance when preparing solutions.

  • Potent CCR4 antagonist for pharmacological research.
  • High purity (99.95%).
  • Solid powder form suitable for dissolution in DMSO or ethanol.
  • Soluble in DMSO or ethanol at 50 mg/mL (~106.9 mM).
  • Powder storage: -20°C for long-term stability; in-solution storage: -80°C recommended.
  • CAS number: 864289-85-0.

Catalog No. 50-002-45134 Supplier Medchemexpress LLC Supplier No. HY10336410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C-021 is a small-molecule CCR4 antagonist (CAS 864289-85-0) provided as a high-purity solid for research use. It is intended for in vitro and in vivo pharmacology studies where CCR4 inhibition is required. Supplied quantity: 10 mg; follow recommended solvent and storage guidance when preparing solutions.

  • Potent CCR4 antagonist for pharmacological research.
  • High purity (99.95%).
  • Solid powder form suitable for dissolution in DMSO or ethanol.
  • Soluble in DMSO or ethanol at 50 mg/mL (~106.9 mM).
  • Powder storage: -20°C for long-term stability; in-solution storage: -80°C recommended.
  • CAS number: 864289-85-0.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.