Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Cyclohexylformamide 99%, Thermo Scientific™

Catalog No. AC153051000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC153051000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC153051000 Supplier Thermo Scientific Chemicals Supplier No. 153051000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 766-93-8
Molecular Formula C7H13NO
Molecular Weight (g/mol) 127.187
InChI Key SWGXDLRCJNEEGZ-UHFFFAOYSA-N
Synonym cyclohexylformamide, formamidocyclohexane, formamide, n-cyclohexyl, n-cyclohexylformamid, n-zyklohexylformamid, n-formylcyclohexylamine, n-cyclohexyl-formamide, unii-x2cfe4sdt6, x2cfe4sdt6, n-cyclohexyl formamide
PubChem CID 13017
ChEBI CHEBI:17945
IUPAC Name N-cyclohexylformamide
SMILES C1CCC(CC1)NC=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 33°C to 39°C
Color White
Infrared Spectrum Authentic
Linear Formula HCONHC6H11
Quantity 100 g
Beilstein 12, II, 11
Formula Weight 127.19
Percent Purity ≥98.5%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material N-Cyclohexylformamide, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.