Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Cyclohexylformamide 98.0+%, TCI America™
SDP

Catalog No. C143625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C143625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C143625G Supplier TCI America Supplier No. C143625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 766-93-8
Molecular Formula C7H13NO
Molecular Weight (g/mol) 127.187
MDL Number MFCD00003828
InChI Key SWGXDLRCJNEEGZ-UHFFFAOYSA-N
Synonym cyclohexylformamide, formamidocyclohexane, formamide, n-cyclohexyl, n-cyclohexylformamid, n-zyklohexylformamid, n-formylcyclohexylamine, n-cyclohexyl-formamide, unii-x2cfe4sdt6, x2cfe4sdt6, n-cyclohexyl formamide
PubChem CID 13017
ChEBI CHEBI:17945
IUPAC Name N-cyclohexylformamide
SMILES C1CCC(CC1)NC=O

Specifications

Color White
Boiling Point 151°C
Quantity 25 g
Formula Weight 127.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material N-Cyclohexylformamide
RTECSNumber LQ1825000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.