Learn More
Medchemexpress LLC N-cyclohexylmaleimide | 1631-25-0 | 99.5% | 179.22 g/mol | C10H13NO2 | 5 G
SDP
Catalog No. 5000299363
Encompass_Preferred
N-cyclohexylmaleimide is a small-molecule maleimide reagent used in conjugation chemistry and organic synthesis. The electrophilic maleimide moiety reacts selectively with thiols and can also react with activated esters (e.g., NHS esters), enabling formation of stable adducts for bioconjugation and labeling. Supplied as a solid for research use.
  • Reactive maleimide group enables rapid conjugation with thiols.
  • Also compatible with activated esters such as NHS esters for diverse coupling strategies.
  • High reported purity (≈99.5%), suitable for research applications.
  • Molecular formula C10H13NO2; molecular weight 179.22 g/mol.
  • Characterized by HNMR and GC; certificate of analysis and safety data available.
  • Available in multiple pack sizes to support small- and medium-scale experiments.
Catalog No. 50-002-99363 Supplier Medchemexpress LLC Supplier No. HYW0165505G
N-cyclohexylmaleimide is a small-molecule maleimide reagent used in conjugation chemistry and organic synthesis. The electrophilic maleimide moiety reacts selectively with thiols and can also react with activated esters (e.g., NHS esters), enabling formation of stable adducts for bioconjugation and labeling. Supplied as a solid for research use.
  • Reactive maleimide group enables rapid conjugation with thiols.
  • Also compatible with activated esters such as NHS esters for diverse coupling strategies.
  • High reported purity (≈99.5%), suitable for research applications.
  • Molecular formula C10H13NO2; molecular weight 179.22 g/mol.
  • Characterized by HNMR and GC; certificate of analysis and safety data available.
  • Available in multiple pack sizes to support small- and medium-scale experiments.