Learn More
Medchemexpress LLC N-cyclohexylmaleimide | 1631-25-0 | MFCD00043904 | 99.5% | 179.22 g/mol | C10H13NO2 | 10 G
SDP
Catalog No. 5000303326
Encompass_Preferred
N-Cyclohexylmaleimide is a maleimide-containing small-molecule reagent used in biochemical research. The maleimide functional group reacts selectively with thiol groups and can participate in reactions with activated esters, enabling bioconjugation and assay development. The material is supplied as a solid with high reported purity and is offered in multiple laboratory pack sizes for research use.
  • Contains a reactive maleimide group suitable for thiol conjugation.
  • High purity for reliable experimental results.
  • Available in multiple laboratory pack sizes.
  • Provided with supporting documentation such as SDS, COA, and data sheet.
  • Suitable for biochemical assays and bioconjugation workflows.
Catalog No. 50-003-03326 Supplier Medchemexpress LLC Supplier No. HYW01655010G
N-Cyclohexylmaleimide is a maleimide-containing small-molecule reagent used in biochemical research. The maleimide functional group reacts selectively with thiol groups and can participate in reactions with activated esters, enabling bioconjugation and assay development. The material is supplied as a solid with high reported purity and is offered in multiple laboratory pack sizes for research use.
  • Contains a reactive maleimide group suitable for thiol conjugation.
  • High purity for reliable experimental results.
  • Available in multiple laboratory pack sizes.
  • Provided with supporting documentation such as SDS, COA, and data sheet.
  • Suitable for biochemical assays and bioconjugation workflows.