Learn More
Medchemexpress LLC N-cyclohexylmaleimide | 1631-25-0 | MFCD00043904 | 99.5% | 179.22 g/mol | C10H13NO2 | 50 G
SDP
Catalog No. 5000301829
Encompass_Preferred
N-Cyclohexylmaleimide is a small-molecule reagent containing a maleimide functional group used for thiol-selective conjugation and crosslinking in chemical biology and synthetic workflows. It is supplied as a high-purity solid suitable for research-scale labeling and crosslinking applications.
  • Reactive maleimide group for selective thiol conjugation.
  • High purity suitable for research applications.
  • Solid form for convenient storage and handling.
  • Compatible with NHS ester chemistries in sequential conjugation workflows.
  • Useful for crosslinking, labeling, and synthesis applications.
Catalog No. 50-003-01829 Supplier Medchemexpress LLC Supplier No. HYW01655050G
Add to Cart
Add to Cart
N-Cyclohexylmaleimide is a small-molecule reagent containing a maleimide functional group used for thiol-selective conjugation and crosslinking in chemical biology and synthetic workflows. It is supplied as a high-purity solid suitable for research-scale labeling and crosslinking applications.
  • Reactive maleimide group for selective thiol conjugation.
  • High purity suitable for research applications.
  • Solid form for convenient storage and handling.
  • Compatible with NHS ester chemistries in sequential conjugation workflows.
  • Useful for crosslinking, labeling, and synthesis applications.