Learn More
Medchemexpress LLC N-Cyclohexyltaurine | 103-47-9 | 99.9% | 25 G
SDP
Catalog No. 5000414150
Encompass_Preferred
CHES (N-Cyclohexyltaurine) is a zwitterionic buffer. It can bind to hemagglutinin (HA) by emulating with sialic acid (SA) and receptor binding site (RBS)-targeting broadly neutralizing antibodies. This product is suitable for various biochemical applications, including fluorescent dye, labeling chemistry, and chemical stain analysis.
  • Zwitterionic buffer
  • Binds to hemagglutinin (HA)
  • Emulates sialic acid (SA) and receptor binding site (RBS)-targeting antibodies
  • Suitable for biochemical assays
  • Used in fluorescent dye applications
  • Applicable in labeling chemistry
  • Useful for chemical stain analysis
Catalog No. 50-004-14150 Supplier Medchemexpress LLC Supplier No. 25GHYD0871
CHES (N-Cyclohexyltaurine) is a zwitterionic buffer. It can bind to hemagglutinin (HA) by emulating with sialic acid (SA) and receptor binding site (RBS)-targeting broadly neutralizing antibodies. This product is suitable for various biochemical applications, including fluorescent dye, labeling chemistry, and chemical stain analysis.
  • Zwitterionic buffer
  • Binds to hemagglutinin (HA)
  • Emulates sialic acid (SA) and receptor binding site (RBS)-targeting antibodies
  • Suitable for biochemical assays
  • Used in fluorescent dye applications
  • Applicable in labeling chemistry
  • Useful for chemical stain analysis