Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(Cyclohexylthio)phthalimide 98.0+%, TCI America™
SDP

Catalog No. C1452500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1452500G 500 g
C145225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1452500G Supplier TCI America Supplier No. C1452500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17796-82-6
Molecular Formula C14H15NO2S
Molecular Weight (g/mol) 261.339
MDL Number MFCD00072247
InChI Key UEZWYKZHXASYJN-UHFFFAOYSA-N
Synonym Cyclohexyl N-Phthalimidyl Sulfide
PubChem CID 28777
IUPAC Name 2-cyclohexylsulfanylisoindole-1,3-dione
SMILES C1CCC(CC1)SN2C(=O)C3=CC=CC=C3C2=O

Specifications

Melting Point 91°C
Color White-Yellow
Quantity 500 g
Formula Weight 261.34
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N-(Cyclohexylthio)phthalimide
EINECSNumber (5)-3358
RTECSNumber TI4290000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.