Learn More
Eurofins Discoverx N-Cyclopentyladenosine, 10 mg. (92-1195)
Supplier:  Eurofins Discoverx 921195
Encompass
N-Cyclopentyladenosine (CPA) is a centrally active synthetic drug compound that acts as an anticonvulsant, as well as a cardiovascular and sleep regulator. Potent and selective agonist of the adenosine receptor ADORA1, with activity for ADORA2A and ADORA3 also (Ki=2.3, 790, and 43 nM, respectively). Provided as 10 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9723