Learn More
AdipoGen N-Dansylethylenediamine (1 g)
SDP
Supplier:  AdipoGen CDXC0068G001
Encompass_Preferred
Chemical. CAS: 35060-08-3. Formula: C14H19N3O2S. MW: 293.38. Dansyl ethylenediamine is a lipophilic fluorophore that can be coupled to coarboxylic acids of proteins and other water-soluble polymers. The primary aliphatic amine of dansyl ethylenediamine can be reversibly coupled to aldehydes and ketones to form a Schiff base, which can be reduced to a generate stable amine derivative by sodium borohydride (NaBH4) or sodium cyanoborohydride (NaCNH3). Spectral data: lambdaEx=335nm; lambdaEm=525nm in methanol. Dansyl ethylenediamine has also shown to inhibit human renal dipeptidase (hrDP) irreversibly in a time-dependent manner.
Catalog No. 50-205-4325