Learn More
Medchemexpress LLC N-DBCO-N-bis(PEG2-C2-acid) | 2110449-00-6 | 96.7% | C33H40N2O10 | 100 MG
SDP
Catalog No. 5000353181
Encompass_Preferred
N-DBCO-N-bis(PEG2-C2-acid) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is a click chemistry reagent, containing a DBCO group that can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains a DBCO group for strain-promoted alkyne-azide cycloaddition (SPAAC) with Azide groups
Catalog No. 50-003-53181 Supplier Medchemexpress LLC Supplier No. HY140543100MG
N-DBCO-N-bis(PEG2-C2-acid) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is a click chemistry reagent, containing a DBCO group that can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains a DBCO group for strain-promoted alkyne-azide cycloaddition (SPAAC) with Azide groups