Learn More
Medchemexpress LLC N-DBCO-N-bis(PEG2-C2-acid) | 2110449-00-6 | 96.7% | C33H40N2O10 | 25 MG
SDP
Catalog No. 5000352838
Encompass_Preferred
N-DBCO-N-bis(PEG2-C2-acid) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules possessing Azide groups. PROTACs comprise two distinct ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets a specific protein, facilitating the selective degradation of target proteins through the intracellular ubiquitin-proteasome system.
  • Peg-based protac linker
  • Click chemistry reagent
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Enables selective degradation of target proteins
Catalog No. 50-003-52838 Supplier Medchemexpress LLC Supplier No. 25MGHY140543
N-DBCO-N-bis(PEG2-C2-acid) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules possessing Azide groups. PROTACs comprise two distinct ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets a specific protein, facilitating the selective degradation of target proteins through the intracellular ubiquitin-proteasome system.
  • Peg-based protac linker
  • Click chemistry reagent
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Enables selective degradation of target proteins