Learn More
Medchemexpress LLC N-DBCO-N-bis(PEG2-C2-NHS ester) | 2128735-29-3 | ≥96.0% | C41H46N4O14 | 50 MG
SDP
Catalog No. 5000356231
Encompass_Preferred
N-DBCO-N-bis(PEG2-C2-NHS ester) is a PEG-based PROTAC linker designed for the synthesis of PROTACs. It functions as a click chemistry reagent due to its DBCO group, which facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules.
  • Functionality: PEG-based PROTAC linker.
  • Application: Used in the synthesis of PROTACs.
  • Mechanism: Contains a DBCO group for strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups.
  • Research Use: For research use only; not sold to patients.
Catalog No. 50-003-56231 Supplier Medchemexpress LLC Supplier No. HY14058550MG
N-DBCO-N-bis(PEG2-C2-NHS ester) is a PEG-based PROTAC linker designed for the synthesis of PROTACs. It functions as a click chemistry reagent due to its DBCO group, which facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules.
  • Functionality: PEG-based PROTAC linker.
  • Application: Used in the synthesis of PROTACs.
  • Mechanism: Contains a DBCO group for strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups.
  • Research Use: For research use only; not sold to patients.