Learn More
Cayman Chemical N-DeacetylcolchIcIn 10mg
SDP
Catalog No. 502986140
Encompass_Preferred
An inhibitor of tubulin polymerization (IC50 = 3 µM for bovine brain tubulin); stimulates tubulin GTPase activity
Catalog No. 50-298-6140 Supplier Cayman Chemical Supplier No. 2169710 MG
An inhibitor of tubulin polymerization (IC50 = 3 µM for bovine brain tubulin); stimulates tubulin GTPase activity