Learn More
TARGETMOL CHEMICALS INC N-decanoyl-L-Homoserine lacton
SDP
Catalog No. 502879035
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. N-decanoyl-L-Homoserine lactone (C10-HSL) is an N-acyl-homoserine lactone (AHL) produced by Pseudomonas cremoris strain ND07.N-decanoyl-L-homoserine lactone inhibits the growth of primary roots of Arabidopsis thaliana.N-decanoyl-L-homoserine lactone treatment caused a transient and immediate increase in free Ca2+ and reactive oxygen species (ROS) concentrations in the root cytoplasm. N-decanoyl-L-homoserine lactone treatment induced transient and immediate increases in cytoplasmic free Ca2+ and reactive oxygen species (ROS) concentrations in Arabidopsis roots, increased mitogen-activated protein kinase 6 (MPK6) activity, and contributed to the production of nitric oxide (NO). Purity 99.81%
Catalog No. 50-287-9035 Supplier TARGETMOL CHEMICALS INC Supplier No. T3773925MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. N-decanoyl-L-Homoserine lactone (C10-HSL) is an N-acyl-homoserine lactone (AHL) produced by Pseudomonas cremoris strain ND07.N-decanoyl-L-homoserine lactone inhibits the growth of primary roots of Arabidopsis thaliana.N-decanoyl-L-homoserine lactone treatment caused a transient and immediate increase in free Ca2+ and reactive oxygen species (ROS) concentrations in the root cytoplasm. N-decanoyl-L-homoserine lactone treatment induced transient and immediate increases in cytoplasmic free Ca2+ and reactive oxygen species (ROS) concentrations in Arabidopsis roots, increased mitogen-activated protein kinase 6 (MPK6) activity, and contributed to the production of nitric oxide (NO). Purity 99.81%