Learn More
Medchemexpress LLC N-Decanoyl-L-homoserine lactone | 177315-87-6 | 98.4% | 255.35 | 100 MG
SDP
Catalog No. 5000348987
Encompass_Preferred
N-Decanoyl-L-homoserine lactone (C10-HSL) is a N-acyl-homoserine lactone (AHL) that has been shown to inhibit primary root growth in Arabidopsis. It triggers an immediate increase in cytosolic free Ca2+ and reactive oxygen species (ROS), enhances mitogen-activated protein kinase 6 (MPK6) activity, and induces nitric oxide (NO) production in Arabidopsis roots. This product is a white to off-white solid with a purity of 98.4%.
  • N-Acyl-homoserine lactone (AHL)
  • Inhibits primary root growth in Arabidopsis
  • Triggers transient increase in cytosolic free Ca2+ and reactive oxygen species (ROS)
  • Increases activity of mitogen-activated protein kinase 6 (MPK6)
  • Induces nitric oxide (NO) production in Arabidopsis roots
  • White to off-white solid appearance
  • Purity of 98.4%
Catalog No. 50-003-48987 Supplier Medchemexpress LLC Supplier No. HY136409100MG
N-Decanoyl-L-homoserine lactone (C10-HSL) is a N-acyl-homoserine lactone (AHL) that has been shown to inhibit primary root growth in Arabidopsis. It triggers an immediate increase in cytosolic free Ca2+ and reactive oxygen species (ROS), enhances mitogen-activated protein kinase 6 (MPK6) activity, and induces nitric oxide (NO) production in Arabidopsis roots. This product is a white to off-white solid with a purity of 98.4%.
  • N-Acyl-homoserine lactone (AHL)
  • Inhibits primary root growth in Arabidopsis
  • Triggers transient increase in cytosolic free Ca2+ and reactive oxygen species (ROS)
  • Increases activity of mitogen-activated protein kinase 6 (MPK6)
  • Induces nitric oxide (NO) production in Arabidopsis roots
  • White to off-white solid appearance
  • Purity of 98.4%