Learn More
Medchemexpress LLC N-decanoyl-L-homoserine lactone | 177315-87-6 | MFCD11113140 | 98.4% | C14H25NO3 | 10MG
SDP
Catalog No. 5000261955
Encompass_Preferred
N-Decanoyl-L-homoserine lactone is an N-acyl-homoserine lactone quorum-sensing signal used in biological research. It modulates microbial and plant signaling pathways and is supplied as a solid or as a DMSO solution for laboratory applications.
  • Small quorum-sensing molecule that modulates bacterial and plant signaling.
  • Reported to inhibit primary root growth and increase cytosolic Ca2+ and reactive oxygen species in Arabidopsis.
  • Induces mitogen-activated protein kinase 6 activity and nitric oxide production in plant roots.
  • White to off-white solid; also available as a 10 mM solution in DMSO.
  • High purity (98.4%) suitable for research applications.
  • Store at 4°C protected from light; in solution, store at -80°C for long-term stability.
Catalog No. 50-002-61955 Supplier Medchemexpress LLC Supplier No. HY13640910MG
N-Decanoyl-L-homoserine lactone is an N-acyl-homoserine lactone quorum-sensing signal used in biological research. It modulates microbial and plant signaling pathways and is supplied as a solid or as a DMSO solution for laboratory applications.
  • Small quorum-sensing molecule that modulates bacterial and plant signaling.
  • Reported to inhibit primary root growth and increase cytosolic Ca2+ and reactive oxygen species in Arabidopsis.
  • Induces mitogen-activated protein kinase 6 activity and nitric oxide production in plant roots.
  • White to off-white solid; also available as a 10 mM solution in DMSO.
  • High purity (98.4%) suitable for research applications.
  • Store at 4°C protected from light; in solution, store at -80°C for long-term stability.