Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-decanoyl-L-homoserine lactone | 177315-87-6 | MFCD11113140 | 99.9% | 255.35 g·mol^-1 | C14H25NO3 | 5 MG
SDP

Catalog No. 5000222270
Encompass_Preferred

N-Decanoyl-L-homoserine lactone is a high-purity analytical standard of N-acyl-homoserine lactone used for method development and quantitative or qualitative analysis in chromatography and mass spectrometry. Supplied as a solid for research and analytical applications.

  • High purity: 99.92%.
  • Molecular formula C14H25NO3 and molecular weight 255.35 g·mol^-1.
  • White to off-white solid appearance.
  • Intended for LC, GC, and MS analytical applications.
  • Available in small vial sizes including 5 mg, 10 mg, 25 mg, and 50 mg.
  • Store under conditions specified in the certificate of analysis; shipped at room temperature in continental US.

Catalog No. 50-002-22270 Supplier Medchemexpress LLC Supplier No. HY136409R5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Decanoyl-L-homoserine lactone is a high-purity analytical standard of N-acyl-homoserine lactone used for method development and quantitative or qualitative analysis in chromatography and mass spectrometry. Supplied as a solid for research and analytical applications.

  • High purity: 99.92%.
  • Molecular formula C14H25NO3 and molecular weight 255.35 g·mol^-1.
  • White to off-white solid appearance.
  • Intended for LC, GC, and MS analytical applications.
  • Available in small vial sizes including 5 mg, 10 mg, 25 mg, and 50 mg.
  • Store under conditions specified in the certificate of analysis; shipped at room temperature in continental US.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.