Learn More
Medchemexpress LLC N-decanoyl-L-homoserine lactone | 177315-87-6 | MFCD11113140 | 99.9% | C14H25NO3 | 10MG
SDP
Catalog No. 5000261956
Encompass_Preferred
N-decanoyl-L-homoserine lactone is an analytical reference standard of the N-acyl homoserine lactone quorum-sensing molecule with a C10 acyl chain. It is supplied as a high-purity solid for use as a calibration or reference material in analytical assays and quorum sensing research. Not for human or clinical use.
  • High purity (>99.9%).
  • Suitable as an analytical reference standard for HPLC and LC-MS.
  • Solid form with molecular weight 255.35 g·mol⁻¹.
  • CAS registry number 177315-87-6 for unambiguous identification.
  • Provided in small analytical package sizes for precise dosing.
  • Intended for research use only.
Catalog No. 50-002-61956 Supplier Medchemexpress LLC Supplier No. HY136409R10MG
N-decanoyl-L-homoserine lactone is an analytical reference standard of the N-acyl homoserine lactone quorum-sensing molecule with a C10 acyl chain. It is supplied as a high-purity solid for use as a calibration or reference material in analytical assays and quorum sensing research. Not for human or clinical use.
  • High purity (>99.9%).
  • Suitable as an analytical reference standard for HPLC and LC-MS.
  • Solid form with molecular weight 255.35 g·mol⁻¹.
  • CAS registry number 177315-87-6 for unambiguous identification.
  • Provided in small analytical package sizes for precise dosing.
  • Intended for research use only.