Learn More
Medchemexpress LLC N-decanoyl-L-homoserine lactone (C10-HSL) | 177315-87-6 | 98.40% | 255.35 | 50 MG
SDP
Catalog No. 5000349462
Encompass_Preferred
N-decanoyl-L-homoserine lactone (C10-HSL) is a N-acyl-homoserine lactone (AHL). It can inhibit primary root growth in Arabidopsis, triggering a transient and immediate increase in cytosolic free Ca2+ and reactive oxygen species (ROS) concentrations. It also increases the activity of mitogen-activated protein kinase 6 (MPK6) and induces nitric oxide (NO) production in Arabidopsis roots.
  • Inhibits primary root growth in Arabidopsis
  • Promotes lateral root formation in Arabidopsis
  • Increases cytosolic Ca2+ concentration in wild-type seedlings
  • Stimulates spore germination of Galactomyces geotrichum
  • For research use only
Catalog No. 50-003-49462 Supplier Medchemexpress LLC Supplier No. HY13640950MG
N-decanoyl-L-homoserine lactone (C10-HSL) is a N-acyl-homoserine lactone (AHL). It can inhibit primary root growth in Arabidopsis, triggering a transient and immediate increase in cytosolic free Ca2+ and reactive oxygen species (ROS) concentrations. It also increases the activity of mitogen-activated protein kinase 6 (MPK6) and induces nitric oxide (NO) production in Arabidopsis roots.
  • Inhibits primary root growth in Arabidopsis
  • Promotes lateral root formation in Arabidopsis
  • Increases cytosolic Ca2+ concentration in wild-type seedlings
  • Stimulates spore germination of Galactomyces geotrichum
  • For research use only