Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Decanoylsarcosine Sodium Salt 98.0+%, TCI America™
SDP

Catalog No. D24895G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D24895G Supplier TCI America Supplier No. D24895G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Anionic surfactant for laboratory research applications.

Chemical Identifiers

CAS 30377-07-2
Molecular Formula C13H24NNaO3
Molecular Weight (g/mol) 265.33
MDL Number MFCD00451533
InChI Key LXXIWMVZOWQLIH-UHFFFAOYSA-M
Synonym (N-Decanoyl-N-methylamino)acetic Acid Sodium Salt, Sodium (N-Decanoyl-N-methylamino)acetate, Sodium N-Decanoylsarcosinate
PubChem CID 44630262
IUPAC Name sodium 2-(N-methyldecanamido)acetate
SMILES [Na+].CCCCCCCCCC(=O)N(C)CC([O-])=O

Specifications

Melting Point 155°C
Color White-Yellow
Quantity 5 g
Formula Weight 265.33
Percent Purity ≥98.0% (T)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material N-Decanoylsarcosine Sodium Salt
EINECSNumber (2)-1226
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.