Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-decyl-N,N-dimethyldecan-1-aminium chloride | 7173-51-5 | MFCD00066262 | 98.0% | 362.08 g/mol | C22H48ClN | 500 MG
SDP

Catalog No. 5000209264
Encompass_Preferred

N-decyl-N,N-dimethyldecan-1-aminium chloride is a dialkyl-quaternary ammonium chloride used as a broad-spectrum antimicrobial agent in laboratory and formulation research.

  • Broad-spectrum bactericidal, virucidal, and fungicidal activity.
  • High purity solid, white to off-white in appearance.
  • CAS number 7173-51-5 and molecular formula C22H48ClN.
  • Soluble in DMSO (100 mg/mL) for in vitro use.
  • Recommended storage: powder at -20°C (long-term) or 4°C (shorter term).
  • Commonly used in antimicrobial testing and formulation studies.

Catalog No. 50-002-09264 Supplier Medchemexpress LLC Supplier No. HYW042181500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-decyl-N,N-dimethyldecan-1-aminium chloride is a dialkyl-quaternary ammonium chloride used as a broad-spectrum antimicrobial agent in laboratory and formulation research.

  • Broad-spectrum bactericidal, virucidal, and fungicidal activity.
  • High purity solid, white to off-white in appearance.
  • CAS number 7173-51-5 and molecular formula C22H48ClN.
  • Soluble in DMSO (100 mg/mL) for in vitro use.
  • Recommended storage: powder at -20°C (long-term) or 4°C (shorter term).
  • Commonly used in antimicrobial testing and formulation studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.