Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-decyl-N,N-dimethyldecan-1-aminium-d6 chloride | 7173-51-5 | 368.11 | C22H42D6ClN | 5 MG
SDP

Catalog No. 5000209207
Encompass_Preferred

N-decyl-N,N-dimethyldecan-1-aminium-d6 chloride is a deuterated quaternary ammonium chloride used as an isotopically labeled internal standard or tracer in analytical and isotopic studies. It is supplied in a small 5 mg package intended for research use.

  • Isotopically labeled (d6) quaternary ammonium chloride.
  • Suitable as an internal standard for analytical methods.
  • Molecular formula C22H42D6ClN.
  • Molecular weight 368.11 g/mol.
  • CAS number 7173-51-5.
  • Supplied as a 5 mg research quantity.

Catalog No. 50-002-09207 Supplier Medchemexpress LLC Supplier No. HYW042181S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-decyl-N,N-dimethyldecan-1-aminium-d6 chloride is a deuterated quaternary ammonium chloride used as an isotopically labeled internal standard or tracer in analytical and isotopic studies. It is supplied in a small 5 mg package intended for research use.

  • Isotopically labeled (d6) quaternary ammonium chloride.
  • Suitable as an internal standard for analytical methods.
  • Molecular formula C22H42D6ClN.
  • Molecular weight 368.11 g/mol.
  • CAS number 7173-51-5.
  • Supplied as a 5 mg research quantity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.