Learn More
Cayman Chemical n-Decyl-β-D-maltoside, Purity: ≥95%, CAS: 82494-09-5, 1 g
SDP
Catalog No. 502936049
Encompass_Preferred
A nonionic surfactant; commonly used to solubilize and stabilize membrane proteins; CMC = 1.8 mM; has been used in the expression of functional recombinant GPCRs
Catalog No. 50-293-6049 Supplier Cayman Chemical Supplier No. 257041 G
A nonionic surfactant; commonly used to solubilize and stabilize membrane proteins; CMC = 1.8 mM; has been used in the expression of functional recombinant GPCRs