Learn More
Strem, An Ascensus Company CAS# 6874-60-8. 5g. n-Decylphosphonic acid, min. 97%. MFCD00015830
SDP
Catalog No. 5090109985
Encompass
CAS# 6874-60-8. 5g. n-Decylphosphonic acid, min. 97%. MFCD00015830. Molecular Weight: 222.26. Molecular Formula: CH3(CH2)9P(O)(OH)2. Color/form: white to off-white pwdr. Strem# 15-0958. http://www.strem.com/catalog/v/15-0958/
Catalog No. 50-901-09985 Supplier Strem, An Ascensus Company Supplier No. 1509585G
CAS# 6874-60-8. 5g. n-Decylphosphonic acid, min. 97%. MFCD00015830. Molecular Weight: 222.26. Molecular Formula: CH3(CH2)9P(O)(OH)2. Color/form: white to off-white pwdr. Strem# 15-0958. http://www.strem.com/catalog/v/15-0958/