Learn More
Medchemexpress LLC N-demethyl ivabradine-d6 hydrochloride | 98.0% | 497.06 g/mol | C26H29D6ClN2O5 | 5 MG
SDP
Catalog No. 5000264609
Encompass_Preferred
N-Demethyl Ivabradine-d6 hydrochloride is the deuterium-labeled N-demethyl metabolite of ivabradine supplied as the hydrochloride salt. It is provided as a solid powder intended for use as an analytical standard or tracer in quantitative NMR, GC-MS, or LC-MS applications.
  • Deuterium-labeled (d6) analogue for use as an internal standard.
  • High purity (98.0%).
  • Provided as a hydrochloride salt in solid powder form.
  • Available in small analytical pack sizes suitable for assay development.
  • Storage stability: powder -20°C (3 years) or 4°C (2 years).
  • Suitable as a tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS.
Catalog No. 50-002-64609 Supplier Medchemexpress LLC Supplier No. HY12778S5MG
N-Demethyl Ivabradine-d6 hydrochloride is the deuterium-labeled N-demethyl metabolite of ivabradine supplied as the hydrochloride salt. It is provided as a solid powder intended for use as an analytical standard or tracer in quantitative NMR, GC-MS, or LC-MS applications.
  • Deuterium-labeled (d6) analogue for use as an internal standard.
  • High purity (98.0%).
  • Provided as a hydrochloride salt in solid powder form.
  • Available in small analytical pack sizes suitable for assay development.
  • Storage stability: powder -20°C (3 years) or 4°C (2 years).
  • Suitable as a tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS.