Learn More
Medchemexpress LLC N-Demethyl Ivabradine hydrochloride | 1246638-08-3 | 99.6% | 1 MG

Supplier: Medchemexpress LLC HY127781MG
N-Demethyl Ivabradine HCl is a metabolite of Ivabradine, which is a specific inhibitor of the funny channel. It is for research use only.
- Specific inhibitor of the funny channel
- White to off-white solid appearance
- Soluble in DMSO
- Store at 4°C, sealed, away from moisture
- In solvent, store at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.