Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-demethyl lincomycin hydrochloride | 14600-41-0 | >95.0% | 428.97 g/mol | C17H33ClN2O6S | 2.5 MG
SDP

Catalog No. 5000285125
Encompass_Preferred

N-demethyl lincomycin hydrochloride is the hydrochloride salt of 4'-propyl-N-demethyllincomycin and is described as an impurity of lincomycin. It is provided for research and analytical use as an impurity/reference standard and is not for human consumption.

  • Hydrochloride salt form suitable for analytical use.
  • Used as an impurity standard for lincomycin impurity profiling.
  • CAS number 14600-41-0 and molecular formula C17H33ClN2O6S.
  • Molecular weight approximately 428.97 g/mol.
  • Typical purity >95.0% (HPLC) for reference use.
  • Provided in small laboratory quantities for analytical workflows.

Catalog No. 50-002-85125 Supplier Medchemexpress LLC Supplier No. HYW5875362.5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-demethyl lincomycin hydrochloride is the hydrochloride salt of 4'-propyl-N-demethyllincomycin and is described as an impurity of lincomycin. It is provided for research and analytical use as an impurity/reference standard and is not for human consumption.

  • Hydrochloride salt form suitable for analytical use.
  • Used as an impurity standard for lincomycin impurity profiling.
  • CAS number 14600-41-0 and molecular formula C17H33ClN2O6S.
  • Molecular weight approximately 428.97 g/mol.
  • Typical purity >95.0% (HPLC) for reference use.
  • Provided in small laboratory quantities for analytical workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.