Learn More
Medchemexpress LLC N-demethyl lincomycin hydrochloride | 14600-41-0 | MFCD00008831 | >95.0% | 428.97 | C17H33ClN2O6S | 1 MG
SDP
Catalog No. 5000283959
Encompass_Preferred
N-demethyl lincomycin hydrochloride is the N-demethylated derivative of lincomycin supplied as the hydrochloride salt for research and analytical applications. It is provided as an impurity reference for impurity profiling and method development in analytical laboratories.
  • Hydrochloride salt form for improved handling and solubility.
  • Intended for use as an analytical standard and impurity reference.
  • Applicable to HPLC and method development workflows.
  • Molecular formula C17H33ClN2O6S and molecular weight 428.97 g/mol.
  • Available in small-scale quantities suitable for analytical testing.
Catalog No. 50-002-83959 Supplier Medchemexpress LLC Supplier No. HYW5875361MG
N-demethyl lincomycin hydrochloride is the N-demethylated derivative of lincomycin supplied as the hydrochloride salt for research and analytical applications. It is provided as an impurity reference for impurity profiling and method development in analytical laboratories.
  • Hydrochloride salt form for improved handling and solubility.
  • Intended for use as an analytical standard and impurity reference.
  • Applicable to HPLC and method development workflows.
  • Molecular formula C17H33ClN2O6S and molecular weight 428.97 g/mol.
  • Available in small-scale quantities suitable for analytical testing.