Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Demethylricinine | 21642-98-8 | 99.9% | C7H6N2O2 | 25 G
SDP

Catalog No. 5000357612
Encompass_Preferred

N-Demethylricinine is a ricinine compound that can be interconverted with ricinine in senescent and green castor plant leaves. It is an α-pyridone alkaloid and its catabolism is linked to the aging process.

  • Appears as a solid.
  • Color is off-white to yellow.
  • Classified as an alkaloid.
  • Derived from plants, Euphorbiaceae, Ricinus communis L.
  • Soluble in DMSO at 50 mg/mL (333.04 mM).
  • Soluble in 10% DMSO, 40% PEG300, 5% Tween-80, 45% saline solution at ≥ 2.5 mg/mL (16.65 mM).
  • Soluble in 10% DMSO, 90% corn oil at ≥ 2.5 mg/mL (16.65 mM).

Catalog No. 50-003-57612 Supplier Medchemexpress LLC Supplier No. 25GHY65008
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Demethylricinine is a ricinine compound that can be interconverted with ricinine in senescent and green castor plant leaves. It is an α-pyridone alkaloid and its catabolism is linked to the aging process.

  • Appears as a solid.
  • Color is off-white to yellow.
  • Classified as an alkaloid.
  • Derived from plants, Euphorbiaceae, Ricinus communis L.
  • Soluble in DMSO at 50 mg/mL (333.04 mM).
  • Soluble in 10% DMSO, 40% PEG300, 5% Tween-80, 45% saline solution at ≥ 2.5 mg/mL (16.65 mM).
  • Soluble in 10% DMSO, 90% corn oil at ≥ 2.5 mg/mL (16.65 mM).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.