Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-desbutyl bupivacaine | 15883-20-2 | MFCD01701244 | 232.32 g/mol | C14H20N2O | 100 G
SDP

Catalog No. 5000210751
Encompass_Preferred

N-Desbutyl bupivacaine is a major metabolite of bupivacaine used as a pharmacological reference compound. It is reported to act as an NMDA receptor inhibitor and an SCN5A antagonist.

  • Cas number: 15883-20-2.
  • Molecular formula: C14H20N2O.
  • Molecular weight: 232.32 g/mol.
  • Available pack sizes: 50 g, 100 g, larger quantities by request.
  • Storage: 4°C, protect from light; in solvent: -80°C (6 months) or -20°C (1 month).
  • Common use: drug metabolite and reference standard in pharmacology and metabolism studies.

Catalog No. 50-002-10751 Supplier Medchemexpress LLC Supplier No. HYW045132100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Desbutyl bupivacaine is a major metabolite of bupivacaine used as a pharmacological reference compound. It is reported to act as an NMDA receptor inhibitor and an SCN5A antagonist.

  • Cas number: 15883-20-2.
  • Molecular formula: C14H20N2O.
  • Molecular weight: 232.32 g/mol.
  • Available pack sizes: 50 g, 100 g, larger quantities by request.
  • Storage: 4°C, protect from light; in solvent: -80°C (6 months) or -20°C (1 month).
  • Common use: drug metabolite and reference standard in pharmacology and metabolism studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.