Learn More
Medchemexpress LLC HY-128554 5mg Medchemexpress, N-Desethyl amodiaquine CAS:79352-78-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1285545MG
Encompass_Preferred
Medchemexpress, HY-128554 5mg N-Desethyl amodiaquine CAS:79352-78-6 N-Desethyl amodiaquine is the major biologically active metabolite of Amodiaquine. N-Desethyl amodiaquine is an antiparasitic agent. IC50 values for strains V1/S and 3D7 are 97 nM and 25 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9752