Learn More
Medchemexpress LLC N-Desethyl amodiaquine dihydrochloride | 79049-30-2 | 99.9% | 1 ML
SDP
Catalog No. 5000342598
Encompass_Preferred
N-Desethyl amodiaquine dihydrochloride is the biologically active metabolite of Amodiaquine. It is an antiparasitic agent that inhibits V1/S and 3D7 strains with IC50 values of 97 nM and 25 nM, respectively.
  • Biologically active metabolite
  • Functions as an antiparasitic agent
  • Inhibits V1/S and 3D7 strains
  • Suitable for malaria research
Catalog No. 50-003-42598 Supplier Medchemexpress LLC Supplier No. HY128554A10MM/1ML
N-Desethyl amodiaquine dihydrochloride is the biologically active metabolite of Amodiaquine. It is an antiparasitic agent that inhibits V1/S and 3D7 strains with IC50 values of 97 nM and 25 nM, respectively.
  • Biologically active metabolite
  • Functions as an antiparasitic agent
  • Inhibits V1/S and 3D7 strains
  • Suitable for malaria research