Learn More
Medchemexpress LLC N-Desethyl amodiaquine dihydrochloride (Standard) | 79049-30-2 | 1 MG
SDP
Catalog No. 5000338266
Encompass_Preferred
N-Desethyl amodiaquine dihydrochloride (Standard) is the analytical standard of N-Desethyl amodiaquine dihydrochloride. This product is intended for research and analytical applications. It is the biologically active metabolite of Amodiaquine and acts as an antiparasitic agent. This compound inhibits Plasmodium falciparum strains V1/S and 3D7 with IC50 values of 97 nM and 25 nM, respectively, and can be utilized for malaria research.
  • Analytical standard
  • Intended for research and analytical applications
  • Biologically active metabolite of Amodiaquine
  • Antiparasitic agent
  • Inhibits Plasmodium falciparum strains V1/S and 3D7
  • Can be used for malaria research
Catalog No. 50-003-38266 Supplier Medchemexpress LLC Supplier No. HY128554S11MG
N-Desethyl amodiaquine dihydrochloride (Standard) is the analytical standard of N-Desethyl amodiaquine dihydrochloride. This product is intended for research and analytical applications. It is the biologically active metabolite of Amodiaquine and acts as an antiparasitic agent. This compound inhibits Plasmodium falciparum strains V1/S and 3D7 with IC50 values of 97 nM and 25 nM, respectively, and can be utilized for malaria research.
  • Analytical standard
  • Intended for research and analytical applications
  • Biologically active metabolite of Amodiaquine
  • Antiparasitic agent
  • Inhibits Plasmodium falciparum strains V1/S and 3D7
  • Can be used for malaria research