Learn More
Cayman Chemical N-desethyl Sunitinib, Purity: ≥95%, CAS: 356068-97-8, 10 mg
SDP
Catalog No. 502987863
Encompass_Preferred
An active metabolite of sunitinib; formed when sunitinib undergoes N-de-ethylation by CYP3A4; is pharmacologically active having similar inhibitory activity to sunitinib
Catalog No. 50-298-7863 Supplier Cayman Chemical Supplier No. 2229210 MG
An active metabolite of sunitinib; formed when sunitinib undergoes N-de-ethylation by CYP3A4; is pharmacologically active having similar inhibitory activity to sunitinib