Learn More
Cayman Chemical N-desethyl SunItInIb 1mg
SDP
Catalog No. 502987918
Encompass_Preferred
An active metabolite of sunitinib; formed when sunitinib undergoes N-de-ethylation by CYP3A4; is pharmacologically active having similar inhibitory activity to sunitinib
Catalog No. 50-298-7918 Supplier Cayman Chemical Supplier No. 222921 MG
An active metabolite of sunitinib; formed when sunitinib undergoes N-de-ethylation by CYP3A4; is pharmacologically active having similar inhibitory activity to sunitinib