Learn More
Medchemexpress LLC HY-10873 50mg , N-Desethyl Sunitinib SU-11662 CAS:356068-97-8 Purity:98%
SDP
Catalog No. 501493848
Encompass_Preferred
Medchemexpress, HY-10873 50mg N-Desethyl Sunitinib SU-11662 CAS:356068-97-8 Formula:C20H23FN4O2 Purity:98% N-Desethyl Sunitinib is a metabolite of sunitinib. Sunitinib is a potent, ATP-competitive VEGFR , PDGFRβ and KIT inhibitor with K i values of 2, 9, 17, 8 and 4 nM for VEGFR -1, -2, -3, PDGFRβ and KIT, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3848 Supplier Medchemexpress LLC Supplier No. HY1087350MG
Medchemexpress, HY-10873 50mg N-Desethyl Sunitinib SU-11662 CAS:356068-97-8 Formula:C20H23FN4O2 Purity:98% N-Desethyl Sunitinib is a metabolite of sunitinib. Sunitinib is a potent, ATP-competitive VEGFR , PDGFRβ and KIT inhibitor with K i values of 2, 9, 17, 8 and 4 nM for VEGFR -1, -2, -3, PDGFRβ and KIT, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.