Learn More
Medchemexpress LLC N-Desethyl Sunitinib | 356068-97-8 | 99.9% | 370.42 | 100 MG
SDP
Catalog No. 5000344658
Encompass_Preferred
N-Desethyl Sunitinib (SU-12662) is a metabolite of Sunitinib. It serves as a good transport substrate for human ABCB1, ABCG2, and murine ABCG2. It is for research use only.
  • Good transport substrate for human ABCB1, ABCG2, and murine ABCG2.
  • Effective against K562 cells.
  • Provided as a solid.
Catalog No. 50-003-44658 Supplier Medchemexpress LLC Supplier No. HY10873S1MG
N-Desethyl Sunitinib (SU-12662) is a metabolite of Sunitinib. It serves as a good transport substrate for human ABCB1, ABCG2, and murine ABCG2. It is for research use only.
  • Good transport substrate for human ABCB1, ABCG2, and murine ABCG2.
  • Effective against K562 cells.
  • Provided as a solid.