Learn More
Medchemexpress LLC N-desethyl sunitinib | 356068-97-8 | 99.9% | 370.42 | 50 MG
SDP
Catalog No. 5000344402
Encompass_Preferred
N-Desethyl Sunitinib (SU-12662) is a metabolite of Sunitinib. It functions as a good transport substrate for human ABCB1, ABCG2, and murine ABCG2. This compound is provided as a solution in DMSO.
  • Metabolite of sunitinib
  • Acts as a good transport substrate for human ABCB1 and ABCG2
  • Functions as a good transport substrate for murine ABCG2
  • Provided in DMSO solution
Catalog No. 50-003-44402 Supplier Medchemexpress LLC Supplier No. HY1087310MM/1ML
N-Desethyl Sunitinib (SU-12662) is a metabolite of Sunitinib. It functions as a good transport substrate for human ABCB1, ABCG2, and murine ABCG2. This compound is provided as a solution in DMSO.
  • Metabolite of sunitinib
  • Acts as a good transport substrate for human ABCB1 and ABCG2
  • Functions as a good transport substrate for murine ABCG2
  • Provided in DMSO solution